missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat REG3G / PAP1B Protein

A DNA sequence encoding the Rattus norvegicus (Rat) REG3G / PAP1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6596-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064420

  • 3260.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_775120.1
REG3G / PAP1B Protein
Research Use Only
Reg3g
Recombinant Protein
E. coli
EDAKEDVPTS RISCPKGSRA YGSYCYALFS VSKSWFDADL ACQKRPSGHL VSVLSGSEAS FVSSLIKSSG NSGQNVWIGL HDPTLGQEPN RGGWEWSNAD VMNYFNWETN PSSVSGSHCG TLTRASGFLR WRENNCISEL PYVCKFKA
Greater than 90% as determined by SDS-PAGE.
24620
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers