missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat S100A9 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) S100A9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6644-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_446039.1 | |
S100A9 Protein | |
Research Use Only | |
S100A9 | |
Recombinant Protein | |
E. coli | |
AAKTGSQLER SISTIINVFH QYSRKYGHPD TLNKAEFKEM VNKDLPNFLK REKRNENLLR DIMEDLDTNQ DNQLSFEECM MLMGKLIFAC HEKLHENNPR GHDHSHGKGC GK | |
Greater than 80% as determined by SDS-PAGE. |
94195 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |