missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat S100A9 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) S100A9, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_446039.1 |
---|---|
Gene ID (Entrez) | 94195 |
Name | S100A9 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024770
|
enQuireBio™
QP6644-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044770
|
enQuireBio™
QP6644-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034770
|
enQuireBio™
QP6644-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054770
|
enQuireBio™
QP6644-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064770
|
enQuireBio™
QP6644-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074770
|
enQuireBio™
QP6644-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_446039.1 | |
S100A9 Protein | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
94195 | |
Research Use Only | |
S100A9 | |
Recombinant Protein | |
E. coli | |
AAKTGSQLER SISTIINVFH QYSRKYGHPD TLNKAEFKEM VNKDLPNFLK REKRNENLLR DIMEDLDTNQ DNQLSFEECM MLMGKLIFAC HEKLHENNPR GHDHSHGKGC GK | |
Greater than 80% as determined by SDS-PAGE. |