missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat VEGF C Protein

A cDNA sequence encoding the VEGF C was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10911-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935548

  • 203.00 € / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

VEGF C Protein
Research Use Only
Measured by its ability to stimulate phosphorylation of the VEGFR-3/FLT-4 receptor in porcine aortic endothelial cells (PAE/FLT -4 cells). The ED50 for this effect is typically 150-300 ng/ml, corresponding to a specific activity of 3.3-6.6KUnits/mg.
Rat
His
The protein was lyophilized from a concentrated (1 mg/ml) solution with BSA.
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Baculovirus
MANUAL
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers