missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat VEGF C Protein
A cDNA sequence encoding the VEGF C was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10911-1mg
Specifications
VEGF C Protein | |
Research Use Only | |
Measured by its ability to stimulate phosphorylation of the VEGFR-3/FLT-4 receptor in porcine aortic endothelial cells (PAE/FLT -4 cells). The ED50 for this effect is typically 150-300 ng/ml, corresponding to a specific activity of 3.3-6.6KUnits/mg. | |
Rat | |
His | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with BSA. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
MANUAL | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |