missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Vesicle-fusing ATPase Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Vesicle-fusing ATPase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8832-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046721

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_068516.1
Vesicle-fusing ATPase Protein
Research Use Only
Nsf
Recombinant Protein
E. coli
QAARCPTDEL SLSNCAVVNE KDYQSGQHVM VRTSPNHKYI FTLRTHPSVV PGCIAFSLPQ RKWAGLSIGQ DIEVALYSFD KAKQCIGTMT IEIDFLQKKN IDSNPYDTDK MAAEFIQQFN HQAFSVGQQL VFSFNDKLFG LLVKDIEAMD PSILKGEPAS GKRQKIEVGL VVGNSQVAFE KAENSSLNLI GKAKTKENRQ SII
Greater than 90% as determined by SDS-PAGE.
60355
10 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers