missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Vesicle-fusing ATPase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Vesicle-fusing ATPase, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_068516.1 |
---|---|
Gene ID (Entrez) | 60355 |
Name | Vesicle-fusing ATPase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036721
|
enQuireBio™
QP8832-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056721
|
enQuireBio™
QP8832-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046721
|
enQuireBio™
QP8832-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066721
|
enQuireBio™
QP8832-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076721
|
enQuireBio™
QP8832-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086721
|
enQuireBio™
QP8832-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_068516.1 | |
Vesicle-fusing ATPase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
60355 | |
Research Use Only | |
Nsf | |
Recombinant Protein | |
E. coli | |
QAARCPTDEL SLSNCAVVNE KDYQSGQHVM VRTSPNHKYI FTLRTHPSVV PGCIAFSLPQ RKWAGLSIGQ DIEVALYSFD KAKQCIGTMT IEIDFLQKKN IDSNPYDTDK MAAEFIQQFN HQAFSVGQQL VFSFNDKLFG LLVKDIEAMD PSILKGEPAS GKRQKIEVGL VVGNSQVAFE KAENSSLNLI GKAKTKENRQ SII | |
Greater than 90% as determined by SDS-PAGE. |