missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rhesus Macaque IL 6 Protein

Product Code. 15984958
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15984958

Brand: enQuireBio™ QP107192ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the IL 6 was constructed and used to recombinantly synthesize the protein.

Specifications

Name IL 6 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Fully biologically active when compared to standard. The ED50 determined by a cell proliferation assay using IL-6-dependent murine 7TD1 cells is less than 0.1ng/ml, corresponding to a specific activity of > 1.0 M 10,000,000 IU/mg.
Product Type Recombinant Protein
Cross Reactivity Rhesus Macaque
Species E. coli
Protein Tag Untagged
Sequence MAPVLPGEDS KNVAAPHSQP LTSSERIDKH IRYILDGISA LRKETCNRSN MCESSKEALA ENNLNLPKMA EKDGCFQSGF NEDTCLVKII TGLLEFEVYL EYLQNRFESS EEQARAVQMS TKVLIQFLQK KAKNLDAITT PEPTTNASLL TKLQAQNQWL QDMTTHLILR SFKEFLQSNL RALRQM
Buffer Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
Purity or Quality Grade Greater than 97.0% as determined by SDS-PAGE and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.