missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rotavirus A Outer capsid glycoprotein VP7 Protein

A DNA sequence encoding the Rotavirus A (strain Human/United Kingdom/ST3/1975 G4-P2A[6]-I1-R1-C1-M1-A1-N1-T1-E1-H1) (RV-A) (Rotavirus A (strain St. Thomas 3)) Rotavirus A Outer capsid glycoprotein VP7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7021-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027080

  • 3590.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Rotavirus A Outer capsid glycoprotein VP7 Protein
Research Use Only
Full Length
Rotavirus
His-B2M
Tris-based buffer, 50% glycerol
1 mg
Not determined.
Recombinant Protein
E. coli
QNYGINLPIT GSMDTAYANS TQDNNFLFST LCLYYPSEAP TQISDTEWKD TLSQLFLTKG WPTGSVYFNE YSNVLEFSID PKLYCDYNVV LIRFVSGEEL DISELADLIL NEWLCNPMDI TLYYYQQTGE ANKWISMGSS CTVKVCPLNT QTLGIGCQTT NTATFETVAD SEKLAIIDVV DSVNHKLNIT STTCTIRNCN KLGPRENVAI IQVGGSNILD ITADPTTSPQ TERMMRVNWK KWWQVFYTVV DYINQIVQVM SKRSRSLDSS SFYYRV
Greater than 85% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers