missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Adapter protein MecA Protein
A DNA sequence encoding the Staphylococcus aureus (strain MW2) Adapter protein MecA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7252-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
WP_001217728.1 | |
S. aureus Adapter protein MecA Protein | |
1 mg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
31213749 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
mecA | |
Recombinant Protein | |
E. coli | |
MRIERVDDTT VKLFITYSDI EARGFSREDL WTNRKRGEEF FWSMMDEINE EEDFVVEGPL WIQVHAFEKG VEVTISKSKN EDMMNMSDDD ATDQFDEQVQ ELLAQTLEGE DQLEELFEQR TKEKEAQGSK RQKSSARKNT RTIIVKFNDL EDVINYAYHS NPITTEFEDL LYMVDGTYYY AVHFDSHVDQ EVINDSYSQL LEFAYPTDRT EVYLNDYAKI IMSHNVTAQV RRYFPETTE |