missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type E Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Pyruvate carboxylase 1, was expressed in the hosts and tags indicated.
385.00 € - 3260.00 €
Specifications
Name | S. aureus Enterotoxin type E Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | entE |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027170
|
enQuireBio™
QP7046-EC-10UG |
10 μg |
385.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047170
|
enQuireBio™
QP7046-EC-200UG |
200 μg |
1535.00 €
200µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037170
|
enQuireBio™
QP7046-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067170
|
enQuireBio™
QP7046-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057170
|
enQuireBio™
QP7046-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017170
|
enQuireBio™
QP7046-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
S. aureus Enterotoxin type E Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
entE | |
Recombinant Protein | |
E. coli | |
SEEINEKDLR KKSELQRNAL SNLRQIYYYN EKAITENKES DDQFLENTLL FKGFFTGHPW YNDLLVDLGS KDATNKYKGK KVDLYGAYYG YQCAGGTPNK TACMYGGVTL HDNNRLTEEK KVPINLWIDG KQTTVPIDKV KTSKKEVTVQ ELDLQARHYL HGKFGLYNSD SFGGKVQRGL IVFHSSEGST VSYDLFDAQG QYPDTLLRIY RDNKTINSEN LHIDLYLYTT | |
Greater than 90% as determined by SDS-PAGE. |