missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Escherichia coli ssPA / Stringent starvation Protein

Product Code. 15956339
Click to view available options
Quantity:
1 mg
250 μg
5 mg
50 μg
500 μg
Unit Size:
1mg
250µg
500µg
50µg
5mg
This item is not returnable. View return policy

Product Code. 15956339

Brand: enQuireBio™ QP552450ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number WP_000676548.1
Name S. aureus Escherichia coli ssPA / Stringent starvation Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol ssPA
Product Type Recombinant Protein
Cross Reactivity S. aureus
Species E. coli
Protein Tag Untagged
Sequence MLPNNDRHQI TDTTNGHYAP VTYIQVEAPT GTFIASGVVV GKDTLLTNKH VVDATHGDPH ALKAFPSAIN QDNYPNGGFT AEQITKYSGE GDLAIVKFSP NEQNKHIGEV VKPATMSNNA ETQVNQNITV TGYPGDKPVA TMWESKGKIT YLKGEAMQYD LSTTGGNSGS PVFNEKNEVI GIHWGGVPNE FNGAVFINEN VRNFLKQNIE DIHFANDDQP NNPDNPDNPN NPDNPNNPDE PNNPDNPNNP DNPDNGDNNN SDNPDAA
Buffer 10 mM sodium phosphate, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.