missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Escherichia coli ssPA / Stringent starvation Protein
Click to view available options
Quantity:
1 mg
250 μg
5 mg
50 μg
500 μg
Unit Size:
1mg
250µg
500µg
50µg
5mg
Specifications
Specifications
| Accession Number | WP_000676548.1 |
| Name | S. aureus Escherichia coli ssPA / Stringent starvation Protein |
| Quantity | 50 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | ssPA |
| Product Type | Recombinant Protein |
| Cross Reactivity | S. aureus |
| Species | E. coli |
| Protein Tag | Untagged |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction