missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Holo-[acyl-carrier-protein] synthase Protein
A DNA sequence encoding the Staphylococcus aureus (strain MRSA252) Holo-[acyl-carrier-protein] synthase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7737-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000581197.1 | |
100 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Holo-[acyl-carrier-protein] synthase Protein | |
Research Use Only | |
acpS | |
Recombinant Protein | |
E. coli | |
MIHGIGVDLI EIDRIKVLYS KQPKLVERIL TKNEQHKFNN FTHEQRKIEF LAGRFATKEA FSKALGTGLG KHVAFNDIDC YNDELGKPKI DYEGFIVHVS ISHTEHYAMS QVVLEKSAF | |
Greater than 90% as determined by SDS-PAGE. |