missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Iron-regulated surface determinant protein 1
A DNA sequence encoding the Staphylococcus aureus (strain USA300 / TCH1516) Iron-regulated surface determinant protein B, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Accession Number | WP_001041586.1 |
---|---|
Name | S. aureus Iron-regulated surface determinant protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | isdB |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006980
|
enQuireBio™
QP6999-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046980
|
enQuireBio™
QP6999-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096970
|
enQuireBio™
QP6999-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026980
|
enQuireBio™
QP6999-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036980
|
enQuireBio™
QP6999-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016980
|
enQuireBio™
QP6999-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
WP_001041586.1 | |
Research Use Only | |
isdB | |
Recombinant Protein | |
E. coli | |
AAEETGGTNT EAQPKTEAVA SPTTTSEKAP ETKPVANAVS VSNKEVEAPT SETKEAKEVK EVKAPKETKE VKPAAKATNN TYPILNQELR EAIKNPAIKD KDHSAPNSRP IDFEMKKKDG TQQFYHYASS VKPARVIFTD SKPEIELGLQ SGQFWRKFEV YEGDKKLPIK LVSYDTVKDY AYIRFSVSNG TKAVKIVSST HFNNKEEKYD YTLMEFAQPI YNSADKFKT | |
Greater than 90% as determined by SDS-PAGE. |
S. aureus Iron-regulated surface determinant protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |