missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Exopolyphosphatase Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Exopolyphosphatase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7169-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047550

  • 3260.00 € / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_012071.1
S. cerevisiae Exopolyphosphatase Protein
Research Use Only
PPX1
Recombinant Protein
E. coli
MSPLRKTVPE FLAHLKSLPI SKIASNDVLT ICVGNESADM DSIASAITYS YCQYIYNEGT YSEEKKKGSF IVPIIDIPRE DLSLRRDVMY VLEKLKIKEE ELFFIEDLKS LKQNVSQGTE LNSYLVDNND TPKNLKNYID NVVGIIDHHF DLQKHLDAEP RIVKVSGSCS SLVFNYWYEK LQGDREVVMN IAPLLMGAIL IDTSNMRRKV EESDKLAIER CQAVLSGAVN EVSAQGLEDS SEFYKEIKSR KNDIKGFSVS DILKKDYKQF NFQGKGHKGL EIGLSSIVKR MSWLFNEHGG EADFVNQCRR FQAERGLDVL VLLTSWRKAG DSHRELVILG DSNVVRELIE RVSDKLQLQL FGGNLDGGVA MFKQLNVEAT RKQVVPYLEE AYSNLEE
Greater than 90% as determined by SDS-PAGE.
856608
1 mg
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers