missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae GTP-binding nuclear protein GSP1 / CNR1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) GTP-binding nuclear protein GSP1/CNR1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7163-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_013396.1 | |
S. cerevisiae GTP-binding nuclear protein GSP1 / CNR1 Protein | |
Research Use Only | |
GSP1 | |
Recombinant Protein | |
Yeast | |
SAPAANGEVP TFKLVLVGDG GTGKTTFVKR HLTGEFEKKY IATIGVEVHP LSFYTNFGEI KFDVWDTAGQ EKFGGLRDGY YINAQCAIIM FDVTSRITYK NVPNWHRDLV RVCENIPIVL CGNKVDVKER KVKAKTITFH RKKNLQYYDI SAKSNYNFEK PFLWLARKLA GNPQLEFVAS PALAPPEVQV DEQLMQQYQQ EMEQATALPL PDEDDADL | |
Greater than 90% as determined by SDS-PAGE. |
851000 | |
500 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |