missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Homocysteine S-methyltransferase 1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Homocysteine S-methyltransferase 1, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_013038.1 |
---|---|
Gene ID (Entrez) | 850664 |
Name | S. cerevisiae Homocysteine S-methyltransferase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026461
|
enQuireBio™
QP8787-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046461
|
enQuireBio™
QP8787-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036461
|
enQuireBio™
QP8787-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056461
|
enQuireBio™
QP8787-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066461
|
enQuireBio™
QP8787-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076461
|
enQuireBio™
QP8787-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_013038.1 | |
S. cerevisiae Homocysteine S-methyltransferase 1 Protein | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |
850664 | |
Research Use Only | |
MHT1 | |
Recombinant Protein | |
E. coli | |
MKRIPIKELI VEHPGKVLIL DGGQGTELEN RGININSPVW SAAPFTSESF WEPSSQERKV VEEMYRDFMI AGANILMTIT YQANFQSISE NTSIKTLAAY KRFLDKIVSF TREFIGEERY LIGSIGPWAA HVSCEYTGDY GPHPENIDYY GFFKPQLENF NQNRDIDLIG FETIPNFHEL KAILSWDEDI ISKPFYIGLS VDDNSLLRDG TTLEEISVHI KGLGNKINKN LLLMGVNCVS FNQSALILKM LHEHLPGMPL LVYPNSGEIY NPKEKTWHRP TNKLDDWETT VKKFVDNGAR IIGGCCRTSP KDIAEIASAV DKYS | |
Greater than 90% as determined by SDS-PAGE. |