missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae IFM1, mitochondrial Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) IFM1, mitochondrial, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_014619.1 |
---|---|
Gene ID (Entrez) | 854135 |
Name | S. cerevisiae IFM1, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
16047560
|
enQuireBio™
QP7172-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067560
|
enQuireBio™
QP7172-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057560
|
enQuireBio™
QP7172-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077560
|
enQuireBio™
QP7172-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087560
|
enQuireBio™
QP7172-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097560
|
enQuireBio™
QP7172-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spécification
NP_014619.1 | |
S. cerevisiae IFM1, mitochondrial Protein | |
Not determined. | |
Full Length | |
S. cerevisiae | |
GST | |
Tris-based buffer, 50% glycerol |
854135 | |
Research Use Only | |
IFM1 | |
Recombinant Protein | |
E. coli | |
KGKRRNQISK KELKPLNFSI PNYISVNKLA NLLNCRVERL IKDLTALGFE NITTTYILSK EYVELILQEY NFALPNLSTS TNLDNVYDEL KSPVNPKLLT KRAPVVTIMG HVDHGKTTII DYLRKSSVVA QEHGGITQHI GAFQITAPKS GKKITFLDTP GHAAFLKMRE RGANITDIIV LVVSVEDSLM PQTLEAIKHA KNSGNEMIIA ITKIDRIPQP KEREKKIEKV INDLIVQGIP VEKIGGDVQV IPISAKTGEN MDLLEESIVL LSEVMDIRAE NSPKTIAEGW IIESQVKKQV GNVATVLVKK GTLQKGKILI CGNTFCKIKN LIDDKGIPIL KATPSYATEV LGWKDVPHVG DEVIQVKSEA IAKKFISKRQ DLIEVQKNSS IVEKLNEERA LAKEQHLNKE LEHENTVQEH EQNTGPKLIN YIIKCDVSGS AEAVSESISS LGNDEVRCNV ISSSVGIPTE SDLKMAQITE STILCFNLGN LPSEVINNRA GIKIKQYNVI YKLIEDVTET LTENLKPIFE KKIVSTVDVR ETFDFRLKKK IIRIAGCKVN NGVIKKNSLV QVVRGPNEDV IFDGKISTLK HNKDDVAEVS KGHECGITFE SGFEGFKPGD KILVYENVRV PRYL | |
Greater than 90% as determined by SDS-PAGE. |