missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant S. marcescens NucA nuclease Protein

Product Code. 18718856
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
This item is not returnable. View return policy

Product Code. 18718856

Brand: R&D Systems™ 10038NA020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Compare to Benzonase™ Nuclease

Measured by its ability to hydrolyze DNA from salmon testes. The specific activity is >130,000 pmol/min/ug, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13717
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 233874471
Molecular Weight (g/mol) M.W. (Observed): 29 kDa, reducing conditions; M.W. (theoretical): 28.9 kDa
Purification Method >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 20 μg
Source E. coli-derived s. marcescens NucA protein Met1-Asn266
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Buffer Supplied as a 0.2 μm filtered solution in Tris and NaCl
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein NucA Nuclease
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.