missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant S. marcescens NucA nuclease Protein
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
Description
Measured by its ability to hydrolyze DNA from salmon testes. The specific activity is >130,000 pmol/min/ug, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P13717 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 233874471 |
| Molecular Weight (g/mol) | M.W. (Observed): 29 kDa, reducing conditions; M.W. (theoretical): 28.9 kDa |
| Purification Method | >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
| Quantity | 20 μg |
| Source | E. coli-derived s. marcescens NucA protein Met1-Asn266 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction