missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Salmon Calcitonin Protein
A cDNA sequence encoding the Calcitonin was constructed and used to recombinantly synthesize the protein.
119.00 € - 1870.00 €
Specifications
Name | Salmon Calcitonin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Salmon |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936648
|
enQuireBio™
QP11261-1MG |
1 mg |
119.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956648
|
enQuireBio™
QP11261-5MG |
5 mg |
481.00 €
5mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946648
|
enQuireBio™
QP11261-25MG |
25 mg |
1870.00 €
25mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Salmon Calcitonin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Salmon | |
Untagged | |
The calcitonin peptide was lyophilized with no additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Calcitonin Acetate (Salmon) has an amino acid sequence of: Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2 | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |