missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Salmonella Typhi S. Typhi HylE Protein

Product Code. 15983389
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 15983389

Brand: enQuireBio™ QP13383100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the S. Typhi HylE was constructed and used to recombinantly synthesize the protein.

Specifications

Name Salmonella Typhi S. Typhi HylE Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Salmonella Typhi
Species E. coli
Protein Tag His
Buffer Lyophilized from 1 mg/ml in 20mM sodium carbonate pH 9.6.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.