missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Salmonella Typhi S. Typhi OMP Protein

A cDNA sequence encoding the S. Typhi OMP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13384-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933399

  • 870.00 € / 500µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Salmonella Typhi S. Typhi OMP Protein
Research Use Only
Recombinant Protein
E. coli
Sterile Filtered solution containing 10mM Tris-HCl, 1mM EDTA and 50mM arginine.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Salmonella Typhi
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers