missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant SARS SARS Core Protein

A cDNA sequence encoding the SARS Core (Amino Acids 1-49) was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13416-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15923499

  • 228.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SARS SARS Core Protein
Research Use Only
Recombinant Protein
E. coli
50mM Tris-HCl, 60mM NaCl and 50% glycerol.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
SARS
Untagged
SARS Core protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers