missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant SARS SARS Mosaic S(M) Protein

A cDNA sequence encoding the SARS Mosaic S(M) was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13421-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15983509

  • 1450.00 € / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SARS SARS Mosaic S(M) Protein
Research Use Only
Recombinant Protein
E. coli
25mM Tris-HCl, 0.4% sarcosyl, 0.25% Triton .–100 and 50% glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
SARS
Untagged
SARS Mosaic protein is >95% pure as determined by 10% PAGE (Coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers