missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 568, Clone: CTK0101 and CTK0102, Proteintech ChromoTek
Description
Nano-Secondary™ alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.
Specifications
Specifications
| Antigen | Recombinant VHH |
| Applications | Immunofluorescence, Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | CTK0101 and CTK0102 |
| Conjugate | Alexa Fluor 568 |
| Host Species | Alpaca |
| Quantity | 10 μL |
| Primary or Secondary | Secondary |
| Target Species | Human, Rabbit |
| Content And Storage | Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?