missing translation for 'onlineSavingsMsg'
Learn More

Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 647, Clone: CTK0101 and CTK0102, Proteintech ChromoTek

Product Code. 17393493
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17393493 10 μL 10µL
17303503 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17393493 Supplier Chromotek Supplier No. srbAF647110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alpaca Monoclonal Antibody

Nano-Secondary™ alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.

Specifications

Antigen Recombinant VHH
Applications Immunofluorescence, Western Blot, Flow Cytometry
Classification Monoclonal
Clone CTK0101 and CTK0102
Conjugate Alexa Fluor 647
Host Species Alpaca
Quantity 10 μL
Primary or Secondary Secondary
Target Species Human, Rabbit
Content And Storage Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.