missing translation for 'onlineSavingsMsg'
Learn More

Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 647, Clone: CTK0105 and CTK0106, Proteintech ChromoTek

Product Code. 17393473
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17393473 10 μL 10µL
17303483 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17393473 Supplier Chromotek Supplier No. sms2bAF647110
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alpaca Monoclonal Antibody

Nano-Secondary™ alpaca anti-mouse IgG2b, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG2b with high affinity & specificity.

Specifications

Antigen Recombinant VHH
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone CTK0105 and CTK0106
Conjugate Alexa Fluor 647
Host Species Alpaca
Quantity 10 μL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.