missing translation for 'onlineSavingsMsg'
Learn More

Recombinant VHH Alpaca, anti-Mouse, Unconjugated, Clone: CTK0103, Proteintech ChromoTek

Product Code. 17373483
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17373483 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17373483 Supplier Chromotek Supplier No. smsG1Cys21500
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Alpaca Monoclonal Antibody

500 μg Nano-Secondary™ for mouse IgG1: alpaca monoclonal Nanobody, Fc-specific VHH, for 2x Cys conjugation, clone [CTK0103].

Specifications

Antigen Recombinant VHH
Applications Immunofluorescence, Western Blot, Flow Cytometry
Clone CTK0103
Host Species Alpaca
Quantity 500 μg
Target Species Mouse
Content And Storage Store at -20°C/-4°F. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.