missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Viral Dengue Virus 3 NS1 Protein
Description
The Recombinant Viral Dengue Virus 3 NS1 Protein is derived from HEK293.
Specifications
Specifications
| Accession Number | Q6YMS4 |
| Characterization | Bioactivity not tested |
| For Use With (Application) | Flow Cytometry |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 41kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell,HEK293-derived viral Dengue Virus 3 NS1 protein Asp774-Ala1125,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10EU per 1μg of the protein by the LAL method. |
| Product Type | Recombinant Protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?