missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Virus Parvovirus B19 VLP VP1/VP2 Protein

Product Code. p-7150847
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15972099

Brand: enQuireBio™ QP129675ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Parvovirus B19 VLP VP1/VP2 was constructed and used to recombinantly synthesize the protein.

Specifications

Name Virus Parvovirus B19 VLP VP1/VP2 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Virus
Species Baculovirus
Protein Tag Untagged
Buffer Parvovirus B19 VLP VP1/VP2 protein solution is supplied in 16mM Sodium phosphate pH 7.4, 120mM NaCl and 20% glycerol.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.