missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus Perth 16/09 Protein
A cDNA sequence encoding the Perth 16/09 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13010-50ug
Additional Details : Weight : 0.01000kg
Specifications
Virus Perth 16/09 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPMQKLPGN DNSTATLCLG HHAVPNGTIV KTITNDQIEV TNATELVQSS STGEICDSPH QILDGKNCTL IDALLGDPQC DGFQNKKWDL FVERSKAYSN CYPYDVPDYA SLRSLVASSG TLEFNNESFN WTGVTQNGTS SACIRRSKNS FFSRLNWLTH LNFKYPALNV TMPNNEQFDK LYIWGVHHPG TDKDQIFLYA QASGRITVST KRSQQTVSPN IGSRPRVRNI PSRISIYWTI VKPGDILLIN STGNLIAPRG YFKIRSGKSS IMRSDAPIGK CNSECITPNG SIPNDKPFQN VNRITYGACP RYVKQNTLKL ATGMRNVPEK QTRHHHHHH | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Virus | |
His | |
The H3N2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |