missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zika NS1 Protein

A cDNA sequence encoding the Zika NS1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP14001-HIS-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965379

  • 700.00 € / 50µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Zika NS1 Protein
Research Use Only
Recombinant Protein
E. coli
MKMASNDATP SNDGAAGLVP EINNEAMALD PVAGAAIAAP LTGQQNIIDP WIMNNFVQAP GGEFTVSPRN SPGEVLLNLE LGPEINPYLA HLARMYNGYA GGFEVQVVLA GNAFTAGKII FAAIPPNFPI ENLSAAQITM CPHVIVDVRQ LEPVNLPMPD VRNNFFHYNQ GSDSKLRLVA MLYTPLRANN SGDDVFTVSC RVLTRPSPDF SFNFLVPPTV ESKTKPFTLP ILTISEMSNS RFPVPIDSLH TSPTENIVVQ CQNGRVTLDG ELMGTTQLLP SQICAFRGVL TRSTSRASDQ ADTATPRLFN YYWHIQLDNL NGTPYDPAED IPGPLGTPDF RGKVFGVASQ RNPDSTTRAH EAKVDTTAGR FTPKLGSLEI STESGDFDQN QPTRFTPVGI GVDHEADFQQ WSLPDYSGQF THNMNLAPAV APNFPGEQLL FFRSQLPSSG GRSNGILDCR VPQEWVQHFY QESAPAQTQV ALVRYVNPDT GRVLFEAKLH KLGFMTIAKN GDSPITVPPN GYFRFESWVN PFYTLAPMGT GNGRRRIQ
Zika NS1 protein is >95% pure as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Zika
His
Zika NS1 protein solution contains PBS, 25mM arginine and 1M urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers