Learn More
Helios Italquartz Rectangular Basins/Dishes, Translucent
Click to view available options
Dimensions (L x W):
60 x 12 mm
68 x 12 mm
Unit Size:
Each
Description
- Translucent
- Rectangular form
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Specifications
| Color | Translucent |
| Material | Quartz |
| Shape | Rectangular |
| Type | Dish |
| Dimensions (L x W) | 60 x 12 mm |
| Length (Metric) | 60/28 mm |
| Width (Metric) | 12 mm |
| Height (Metric) | 12 mm |