Learn More
3M™ Red SMS Based Fabric Category III Protective Coveralls
Product Code. 11923764 Shop All 3M Products
Change view
Click to view available options
Size: XX-Large
Product Code.
11923764 Size: XX-Large
1 options
Product Code. 11923764 Supplier 3M™ Supplier No. 7000089629
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Product Type General Purpose Coveralls
Gender Unisex
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel
Seam Type Serged
Sterility Non-sterile
Protection Class Category III
Height (Metric) 186 to 194 cm