missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Redox Solution, 430mV Pt/Calomel, 470mV Pt/Ag/AgCl

Product Code. 10664471 Shop All SI Analytics Products
Change view
Click to view available options
Quantity:
1000 mL
60 Ampules/20mL
Unit Size:
1000mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10664471 1000 mL 1000mL
10340641 60 Ampules/20mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10664471 Lieferant SI Analytics™ Lieferanten-Nr. 285138184
 Restricted Item
Estimated Shipment: 06-01-2027
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SI Analytics™ Redox Solution, 430mV Pt/Calomel, 470mV Pt/Ag/AgCl is designed to be used in redox tests.

  • GHS07: Skin Irrit. 2 H315 Causes skin irritation, Eye Irrit. 2 H319 Causes serious eye irritation
  • Precautionary statements: P280 Wear protective gloves / eye protection / face protection. P264 Wash thoroughly after handling. P305+P351+P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P321 Specific treatment (see on this label). P332+P313 If skin irritation occurs: Get medical advice/attention. P337+P313 If eye irritation persists: Get medical advice/attention. P302+P352 IF ON SKIN: Wash with plenty of water.
  • Ingredients with limit values that require monitoring at the workplace: 7664-93-9 sulphuric acid
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 102°C
Color Light Yellow
Conductivity 430 Pt/Kalomel, 470 Pt/Ag/Agcl
Filling Solution Type Redox
Odor Odorless
Packaging DURAN™ Bottle
pH Range -0.1
Quantity 1000 mL
Solubility Fully miscible in water
Vapor Pressure 23 hPa
compliance-icons compliance-icons
Product Identifier
  • Redox testing solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.