Learn More
Hach Reference Electrode XR110, Hg Free
Product Code. 17809256
Change view
Click to view available options
For Use With (Equipment):
Ion Selective Electrode
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17809256 Ion Selective Electrode Each
1 options
Product Code. 17809256 Supplier Hach Supplier No. B20B112
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Radiometer analytical XR110 reference electrode with 30mL KCl 3M refill solution and operating instruction.
  • The XR110 Reference electrode has a glass body, a red rod Ag/AgCl reference technology system, refillable electrolyte (KCl 3M), a ground joint, and a porous pin junction
  • The XR110 reference probe is recommended for voltammetry applications and educational purposes
  • The XR110 is a screw cap electrode and requires an external connection cable for connection to a meter. (Replace Obsolete #B20B110)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ion Selective Electrode
Type Refill Reference Electrode