Learn More
Contec Refibe Quiltec Mop Head
Product Code. 31777565 Shop All Contec Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
31777565 Non-sterile Pack of 40
31777560 Sterile Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31777565 Supplier Contec Supplier No. RFQT0100
  • ONLINE EXCLUSIVE PRICE
    171.23 €
    126.38 € / Pack of 40
    Save 44.85 €
    26% Off
View all promotions
Estimated Shipment: 28-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Quicktask ReFIBE Mop Heads are ideal for even the highest grade of cleanroom as they generate very small numbers of particle and fibres.
  • QuickTask ReFIBE Mop Heads are manufactured from 100% recycled 2-ply polyester around a recycled polyester core
  • These mop heads have been specifically designed for use with Contecs Mop Saturation Systems. With high absorbency the mops are suitable for single use saturation, giving a surface coverage up to approximately 20 sqm.
  • Also suitable for the application of disinfectants and removal of light soil, residues and biofilms.
  • Designed for single use, the mop heads are compatible with a range of chemicals and disinfectants.
  • Made from 100% recycled polyester materials
  • 100% recycled 2-ply polyester mop heads
  • High sorbency
  • Available sterile
  • Suitable for hands free mop application with appropriate hardware
  • Double bagged
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mop Pad
Sterility Non-sterile
Autoclavable Not Autoclavable
Color-coded No
Material Polyester