missing translation for 'onlineSavingsMsg'
Learn More

Labcon Refillable Racks for Labcon Tips

Product Code. 16621963 Shop All Labcon Products
Change view
Click to view available options
Holds:
Tips (10 to 1250μL)
Tips (Labcon 10μL)
Unit Size:
Pack of 10
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
16621963 Tips (10 to 1250μL) Pack of 10
16681963 Tips (Labcon 10μL) Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16621963 Supplier Labcon Supplier No. 12109600069
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Eclipse™ pipette tip refills

  • Pipette tip rack
  • Refillable
  • With hinged clear lid
  • Made in USA

Specifications

Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Autoclavable Autoclavable
Color Purple
Holds Tips (10 to 1250μL)
Material Polypropylene
Positions Holds one refill platform of 96 tips
Sterility Non-sterile
For Use With (Application) Liquid Handling
Packaging 10 racks / Pk./10 Pk. / case
Quantity 10 racks / Pk.
Shipping Condition RT
Size 10-1250 μL tips
Product Type Empty Refill Rack with Lid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.