Learn More
Bellingham Stanley™ Eclipse Professional Grade Optical Hand-Held Refractometer
Hand-held refractometers are low-cost, portable instruments used for “on the spot” determination of Refractive Index (RI) of liquids or the concentration of solutions. Offering a choice of scales, Eclipse refractometers can be used in a multitude of applications across many and varied industries.
Brand:  Bellingham Stanley™ 45-46
Code : 04
Additional Details : Weight : 0.01000kg
Product Code. 12343529
  • 367.82 € / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
  • Range: antifreeze - % Ethylene Glycol 0 to 60; % Propylene Glocol 0 to 60
  • Simple zero adjust with lock
  • Shaded field of view for distinction
  • High precision, clear scale
  • Easy-clean prism
  • Unique sample “dribble” feature
  • Low weight
  • Anti-roll supports
  • Push-on prism flap
Specifications
Specifications
Handheld Refractometer
2.5
Measurement of heat transfer fluids in air conditioning plants, heat exchangers, pasteurisers, and fluid based radiator systems, automobile manufacturer cooling system loading, and more
0 to 60 Brix
Antifreeze - % concentration by vol., ethylene glycol and propylene glycol
Certificate of calibration, foam carry case and instruction manual