Learn More
Labcold™ Refrigerator RPFG05043 advanced, underbench, glass door with lock
Product Code. 12948624
Change view
Click to view available options
alarm, 2 shelves, stainless steel exterior fan assisted 230V 50/60Hz 2°C to 8°C 600mm x 620mm x 820mm (w x d x h), 150L Labcold
Product Code.
12948624 alarm, 2 shelves, stainless steel exterior fan assisted 230V 50/60Hz 2°C to 8°C 600mm x 620mm x 820mm (w x d x h), 150L Labcold
1 options
Product Code. 12948624 Supplier Labcold™ Supplier No. RPFG05043
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Microprocessor controlled temperature regulation for accuracy and stability
  • Temperature range +2°C to +8°C
  • Fan circulated interior
  • Stainless steel construction
  • Door locks fitted as standard
  • Digital temperature display with audio alarm
  • Heavy duty lockable castors on larger models
  • Optimised air circulation
12mm access port also available as a factory fitted option, please enquire.