missing translation for 'onlineSavingsMsg'
Learn More

Regadenoson, MedChemExpress

Product Code. 16444197
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444197

Brand: Medchem Express HYA0168

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Regadenoson (CVT-3146) is a potent and selective A2A adenosine receptor agonist, with Kis of 290 and 1120 nM for rat and pig adenosine A2A receptor, respectively.

Regadenoson is selective for the adenosine A2A receptor over adenosine A1 and A2B receptors, and shows 13-fold selectivity over the human adenosine A1 receptor. Regadenoson is a vasodilator stress agent has shifted the landscape of vasodilator myocardial perfusion imaging. Regadenoson increases blood-brain barrier (BBB) permeability in rodents.

Specifications

Chemical Name or Material Regadenoson
CAS 313348-27-5
Color White
Purity Grade Notes Research
Molecular Formula C15H18N8O5
Quantity 50 mg
Synonym CVT-3146
Solubility Information DMSO : 50 mg/mL (128.09 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=C(N4N=CC(C(NC)=O)=C4)N=C3N)O1)O)O
Molecular Weight (g/mol) 390.35
Formula Weight 390.35
Percent Purity 98.35%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.