Learn More
WTW™ Regeneration Set for WTW™ Oxygen Sensors CellOx 325, TA 197 Oxi, ConOx
Product Code. 10245681
Change view
Click to view available options
For Use With (Equipment):
Sensor CellOx 325 ConOx, TA 197 Oxi and MPP 350
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10245681 Sensor CellOx 325 ConOx, TA 197 Oxi and MPP 350 Each
1 options
Product Code. 10245681 Supplier WTW™ Supplier No. 202706
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Membrane-covered galvanic dissolved oxygen sensors to measure in water and waterous solutions.
  • Regeneration Set for WTW™ Oxygen Sensors CellOx 325, TA 197 Oxi, ConOx
  • Membrane-covered galvanic dissolved oxygen sensors to measure in water and waterous solutions - low-maintenance and no polarization time, required
  • For instant measurement
  • Easy air calibration with water vapor saturated air in the calibration vessel
  • User friendly and long operation time
TRUSTED_SUSTAINABILITY
Specifications
Description Regeneration set
Includes Electrolyte, cleaning solution, 3 interchangeable heads foil diaphragm and grinding
For Use With (Equipment) Sensor CellOx 325 ConOx, TA 197 Oxi and MPP 350