missing translation for 'onlineSavingsMsg'
Learn More

Regorafenib-d3, TRC

Product Code. 30417783
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417783 10 mg 10mg
30424388 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30417783 Supplier Toronto Research Chemicals Supplier No. TRCR14300210MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Regorafenib-d3
CAS 1255386-16-3
Purity Grade Notes HPLC
Molecular Formula C21H12D3ClF4N4O3
Quantity 10 mg
Synonym 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]-3-fluorophenoxy]-N-(methyl-d3)-2-pyridinecarboxamide
InChI Formula InChI=1S/C21H15ClF4N4O3/c1-27-19(31)18-10-13(6-7-28-18)33-12-3-5-17(16(23)9-12)30-20(32)29-11-2-4-15(22)14(8-11)21(24,25)26/h2-10H,1H3,(H,27,31)(H2,29,30,32)/i1D3
SMILES O=C(NC1=CC=C(Cl)C(C(F)(F)F)=C1)NC2=C(F)C=C(OC3=CC=NC(C(NC([2H])([2H])[2H])=O)=C3)C=C2
Molecular Weight (g/mol) 485.83
Formula Weight 485.0957
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.