missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Regular Fetal Bovine Serum
BIOPHARMA

Product Code. 15343681
Change view
Click to view available options
Quantity:
500 mL
Product Type:
Fetal Bovine Serum
Heat Inactivated:
No
pH:
7.6 ±0.8
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Heat Inactivated pH Unit Size
15343681 500 mL Fetal Bovine Serum No 7.6 ±0.8 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15343681 Supplier Corning™ Supplier No. 35010CV
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.

  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Regular FBS is sourced from herds in USDA approved countries (ex: Mexico, Honduras, Costa Rica, Nicaragua, etc.)

Beställningsinfo

Shipped in frozen form

TRUSTED_SUSTAINABILITY

Specifikationer

Certifications/Compliance cGMP
Color Amber Yellow to Reddish
Content And Storage -25° to -15°C
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Bovine Fetus
Grade Regular
Endotoxin Concentration ≤20EU/ mL
For Use With (Application) Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only.
Heat Inactivated No
Hemoglobin Concentration <30 mg/dL
Osmolality 315 ± 35mOsm/kg
pH 7.6 ±0.8
Protein Content 30 to 50g/L
USDA Safety Testing Included USDA Inspected
Visa mer Visa mindre

For research use only.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.