missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Regular Fetal Bovine Serum
BIOPHARMAGREENER_CHOICE

Product Code. p-4255452
Click to view available options
Quantity:
500 mL
Product Type:
Fetal Bovine Serum
Heat Inactivated:
No
pH:
7.6 ±0.8
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15343681

missing translation for 'mfr': Corning™ 35010CV

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.

  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Regular FBS is sourced from herds in USDA approved countries (ex: Mexico, Honduras, Costa Rica, Nicaragua, etc.)

Order Info

Shipped in frozen form

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance cGMP
Color Amber Yellow to Reddish
Content And Storage -25° to -15°C
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Bovine Fetus
Grade Regular
Endotoxin Concentration ≤20EU/ mL
For Use With (Application) Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only.
Heat Inactivated No
Hemoglobin Concentration <30 mg/dL
Osmolality 315 ± 35mOsm/kg
pH 7.6 ±0.8
Protein Content 30 to 50g/L
USDA Safety Testing Included USDA Inspected
Show More Show Less

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.