missing translation for 'onlineSavingsMsg'
Learn More

Reinecke Salt, Honeywell™

Product Code. 15628070 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15628070

Brand: Honeywell Research Chemicals R412625G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for use as a reagent for choline determination

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13573-16-5
MDL Number MFCD00066638

Specifications

Melting Point 268°C to 272°C
Physical Form Solid
Quantity 25 g
Linear Formula NH4[Cr(NH3)2(SCN)4]
UN Number NONH for all modes of transport
Formula Weight 336.43g/mol
Packaging Poly bottle
Chemical Name or Material Reinecke salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.