missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Reinforced Large Volume Container

Product Code. 17958891
Click to view available options
Capacity (Metric):
170 L
220 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17958891

Brand: Gilac™ G145521

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Reinforced Large Volume Container has excellent resistance for intensive use and extreme handling conditions.

  • Excellent mechanical resistance: made from high density polyethylene for increased shock resistance.
  • The durable, thick walls mean no distortion and warping.
  • Limited gaps and overlapping, equipped with edges so easy to handle.
  • Volume in litres is equivalent to the container filled to the brim.

Specifications

Type Storage Tote
Material HDPE
Color White
Shape Rectangular
Capacity (English) 44.9 gal.
Capacity (Metric) 170 L
Height (English) 2.3 ft.
Height (Metric) 700 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.