missing translation for 'onlineSavingsMsg'
Learn More

rel-N-[5-[4-[5-[[(2R,6S)-2,6-Dimethyl-4-morpholinyl]methyl]-2-oxazolyl]-1H-indazol-6-yl]-2-methoxy-3-pyridinyl]methanesulfonamide, TRC

Product Code. 30242994
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30242994 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30242994 Supplier Toronto Research Chemicals Supplier No. TRCG7974301MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1254036-66-2
Molecular Formula C24H28N6O5S
Quantity 1 mg
Synonym GSK 2292767;
InChI Formula InChI=1S/C24H28N6O5S/c1-14-11-30(12-15(2)34-14)13-18-9-26-23(35-18)19-5-16(6-21-20(19)10-27-28-21)17-7-22(29-36(4,31)32)24(33-3)25-8-17/h5-10,14-15,29H,11-13H2,1-4H3,(H,27,28)/t14-,15+
SMILES COc1ncc(cc1NS(=O)(=O)C)c2cc(c3oc(CN4C[C@@H](C)O[C@@H](C)C4)cn3)c5cn[nH]c5c2
IUPAC Name N-[5-[4-[5-[[(2R,6S)-2,6-dimethylmorpholin-4-yl]methyl]-1,3-oxazol-2-yl]-1H-indazol-6-yl]-2-methoxypyridin-3-yl]methanesulfonamide
Molecular Weight (g/mol) 512.58
Formula Weight 512.1842
Chemical Name or Material rel-N-(5-(4-(5-(((2R,6S)-2,6-Dimethyl-4-morpholinyl)methyl)-2-oxazolyl)-1H-indazol-6-yl)-2-methoxy-3-pyridinyl)methanesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.